missing translation for 'onlineSavingsMsg'
Learn More

CRIK Mouse, Unlabeled, Clone: 6, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 611376

 View more versions of this product

Product Code. 15895899

  • ONLINE EXCLUSIVE PRICE
    £198.00
    £190.20 / 1 set
    Save £7.80
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Citron was identified by its interactions with activated Rho and Rac. It contains a zinc finger (ZF) region, a pleckstrin homology (PH) domain, a citron/MRCK/Gek (CMG) domain, and a coiled-coil domain with a Rho-Rac binding region (RRB). Citron binds to PSD-95 via its C-terminal tSXV motif and the PDZ domain on PSD-95. This association occurs at post synaptic densities where citron-PSD-95 may provide a specific link between the Rho signaling cascade and the synaptic NMDA receptor complex. Citron Rho-interacting kinase (CRIK) and CRIK short kinase (CRIK-SK) are Ser/Thr kinase isoforms of the myotonic dystrophy kinase family. CRIK contains full-length citron, with an N-terminal kinase domain. CRIK-SK is a splice variant that contains only the kinase domain. CRIK co-localizes with actin in keratinocytes and co-expression of CRIK and Rho increases CRIK kinase activity two-fold. In addition, CRIK mutants cause abnormal contraction during cytokinesis. CRIK is expressed highest in testis, but is also present in brain, spleen, lung, and kidney. Thus, CRIK is a Ser/Thr kinase that functions downstream of Rho during events such as cytokinesis and synaptic transmission.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CRIK
Monoclonal
250μg/mL
Serine/Threonine Protein Kinase 21
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
6
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Mouse CRIK aa. 1420-1612
50 μg
Cell Biology
Human, Mouse, Rat
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.