Learn More
CD62L Mouse anti-Human, V450, Clone: DREG 56, BD
Mouse Monoclonal Antibody
£154.24 - £306.02
Specifications
| Antigen | CD62L |
|---|---|
| Clone | DREG 56 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Violet Fluor 450 (V450) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15870199
|
BD Biosciences
560441 |
30 Tests |
£161.00 £154.24 / Each Save £6.76 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15860199
|
BD Biosciences
560440 |
120 Tests |
£310.00 £306.02 / Each Save £3.98 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Reacts with a 76-95kDa glycoprotein referred to as L-selectin or LECAM-1. CD62L is expressed on neutrophils, monocytes, T- and B-lymphocyte subsets and NK cells. Dreg-56 antibody recognizes the same antigen as LAM-1, and specifically inhibits >90% of binding of human lymphocytes to high endothelial venules (HEV) in frozen sections of peripheral, but not mucosal lymphoid tissue. It thus defines L-selectin as a human lymphocyte homing receptor for peripheral lymph node HEV. The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.
Host Species: Mouse
Clone: DREG-56
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Specifications
| CD62L | |
| Flow Cytometry | |
| Violet Fluor 450 (V450) | |
| RUO | |
| Human | |
| Supernatant from PMA-activated Human Peripheral Blood Leukocytes | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| DREG 56 | |
| Monoclonal | |
| Mouse | |
| Immunology | |
| Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified | |
| SELL; L-selectin; LSEL; LAM-1; LECAM-1; LEU8; LNHR; MEL-14; PLNHR; TQ-1 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.