Learn More
CD62L Mouse anti-Human, V450, Clone: DREG 56, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 560440
Description
Reacts with a 76-95kDa glycoprotein referred to as L-selectin or LECAM-1. CD62L is expressed on neutrophils, monocytes, T- and B-lymphocyte subsets and NK cells. Dreg-56 antibody recognizes the same antigen as LAM-1, and specifically inhibits >90% of binding of human lymphocytes to high endothelial venules (HEV) in frozen sections of peripheral, but not mucosal lymphoid tissue. It thus defines L-selectin as a human lymphocyte homing receptor for peripheral lymph node HEV. The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.
Host Species: Mouse
Clone: DREG-56
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Specifications
| CD62L | |
| Monoclonal | |
| Violet Fluor 450 (V450) | |
| Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
| Supernatant from PMA-activated Human Peripheral Blood Leukocytes | |
| 120 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| DREG 56 | |
| SELL; L-selectin; LSEL; LAM-1; LECAM-1; LEU8; LNHR; MEL-14; PLNHR; TQ-1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.