Learn More
CD3 Mouse anti-Human, APC-H7, Clone: SK7, BD
Mouse Monoclonal Antibody
£157.48 - £331.43
Specifications
| Antigen | CD3 |
|---|---|
| Clone | SK7 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | APC-H7 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15680457
|
BD Biosciences
560275 |
25 Tests |
£164.00 £157.48 / Each Save £6.52 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15849988
|
BD Biosciences
560176 |
100 Tests |
£621.00 £331.43 / Each Save £289.57 46% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD3 reacts with ε chain of the CD3 antigen/T-cell antigen receptor (TCR) complex. This complex is composed of at least six proteins that range in molecular weight from 20 to 30kDa. The antigen recognized by CD3 antibodies is noncovalently associated with either α/β or γ/δ TCR (70 to 90kDa). The CD3 antigen is present on 61% to 85% of normal peripheral blood lymphocytes 60% to 85% of thymocytes and on Purkinje cells in cerebellum. This antibody has a Mitogenic effect on most peripheral blood T lymphocytes, provided appropriate functional monocytes are present.
Host Species: Mouse
Clone: SK7
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Specifications
| CD3 | |
| Flow Cytometry | |
| APC-H7 | |
| RUO | |
| Human | |
| Human Thymocytes | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| SK7 | |
| Monoclonal | |
| Mouse | |
| Immunology | |
| Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified | |
| CD3-epsilon; CD3E; Leu4; T-cell surface antigen T3/Leu-4 epsilon chain; T3E |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.