missing translation for 'onlineSavingsMsg'
Learn More

CD3 Mouse anti-Human, APC-H7, Clone: SK7, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 560176

 View more versions of this product

Product Code. 15849988

  • ONLINE EXCLUSIVE PRICE
    £621.00
    £331.43 / Each
    Save £289.57
    46% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CD3 reacts with ε chain of the CD3 antigen/T-cell antigen receptor (TCR) complex. This complex is composed of at least six proteins that range in molecular weight from 20 to 30kDa. The antigen recognized by CD3 antibodies is noncovalently associated with either α/β or γ/δ TCR (70 to 90kDa). The CD3 antigen is present on 61% to 85% of normal peripheral blood lymphocytes 60% to 85% of thymocytes and on Purkinje cells in cerebellum. This antibody has a Mitogenic effect on most peripheral blood T lymphocytes, provided appropriate functional monocytes are present.

Host Species: Mouse
Clone: SK7
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD3
Monoclonal
APC-H7
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Human Thymocytes
100 Tests
Immunology
Human
IgG1 κ
Flow Cytometry
SK7
CD3-epsilon; CD3E; Leu4; T-cell surface antigen T3/Leu-4 epsilon chain; T3E
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.