Learn More
CD22 Mouse anti-Human, FITC, Clone: HIB22, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555424
Description
The HIB22 monoclonal antibody specifically binds to CD22. CD22 is a 130-140 kDa glycosylated type I integral membrane protein present on the surface of mature B cells. CD22 is expressed in the cytoplasm of virtually all B cells except plasma cells. CD45RO antigen on T cells and CD75 antigen on B cells have been identified as ligands for CD22. CD22 has been reported to participate in B-cell activation and also as an adhesion molecule. Although the immunobiology of this antigen has not been fully elucidated, reports indicate that ligation of CD22 induces constitutive internalization of the molecule followed by complete degradation.
Specifications
| CD22 | |
| Monoclonal | |
| FITC | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| HIB22 | |
| BL-CAM; Siglec-2; Bgp135; Lyb8; LPAP | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.