missing translation for 'onlineSavingsMsg'
Learn More

CD22 Mouse anti-Human, FITC, Clone: HIB22, BD

Mouse Monoclonal Antibody

£117.51 - £226.83

Specifications

Antigen CD22
Clone HIB22
Applications Flow Cytometry
Classification Monoclonal
Conjugate FITC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15861319
View Documents
BD Biosciences
561771
25 Tests
ONLINE EXCLUSIVE PRICE
£122.00
£117.51 / Each
Save £4.49
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15808158
View Documents
BD Biosciences
555424
100 Tests
ONLINE EXCLUSIVE PRICE
£236.00
£226.83 / Each
Save £9.17
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The HIB22 monoclonal antibody specifically binds to CD22. CD22 is a 130-140 kDa glycosylated type I integral membrane protein present on the surface of mature B cells. CD22 is expressed in the cytoplasm of virtually all B cells except plasma cells. CD45RO antigen on T cells and CD75 antigen on B cells have been identified as ligands for CD22. CD22 has been reported to participate in B-cell activation and also as an adhesion molecule. Although the immunobiology of this antigen has not been fully elucidated, reports indicate that ligation of CD22 induces constitutive internalization of the molecule followed by complete degradation.

Specifications

Specifications

CD22
Flow Cytometry
FITC
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
HIB22
Monoclonal
Mouse
Human
IgG1 κ
Affinity Purified
BL-CAM; Siglec-2; Bgp135; Lyb8; LPAP
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.