Learn More
CD184 Mouse anti-Human, APC, Clone: 12G5, BD
Mouse Monoclonal Antibody
£298.00 - £473.00
Specifications
Antigen | CD184 |
---|---|
Clone | 12G5 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | APC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15800619
|
BD Biosciences
560936 |
25 Tests |
£298.00
Each |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10107913
|
BD Biosciences
555976 |
100 Tests |
£473.00
Each |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Reacts with fusin (CXCR4), a seven-transmembrane domain, G-protein-linked glycoprotein. Fusin is expressed on a wide variety of CD4+ and CD4- cells including those from spleen, brain and lung. It has been shown to function as a cofactor for infection with T-cell tropic strains of HIV-1 and also as a receptor for CD4-independent infection. 12G5 antibody has been reported to block CD4-independent infection by HIV-2 and CD4-dependent infection by some T-cell tropic isolates of HIV-1.
Host Species: Mouse
Clone: 12G5
Isotype: IgG2a κ
Species Reactivity: Human
Flow Cytometry
Specifications
CD184 | |
Flow Cytometry | |
APC | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG2a κ | |
Affinity Purified | |
CXCR4; Fusin; SDF-1 receptor; LAP3; LCR1; LESTR; NPYY3R; NPY3R; WHIM; HM89 |
12G5 | |
Monoclonal | |
Mouse | |
Human | |
SIVmac variant CP-MAC-infected Sup-T1 cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.