missing translation for 'onlineSavingsMsg'
Learn More

CD184 Mouse anti-Human, APC, Clone: 12G5, BD

Product Code. 15800619
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15800619 25 Tests Each
10107913 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15800619 Supplier BD Biosciences Supplier No. 560936

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Reacts with fusin (CXCR4), a seven-transmembrane domain, G-protein-linked glycoprotein. Fusin is expressed on a wide variety of CD4+ and CD4- cells including those from spleen, brain and lung. It has been shown to function as a cofactor for infection with T-cell tropic strains of HIV-1 and also as a receptor for CD4-independent infection. 12G5 antibody has been reported to block CD4-independent infection by HIV-2 and CD4-dependent infection by some T-cell tropic isolates of HIV-1.

Host Species: Mouse
Clone: 12G5
Isotype: IgG2a κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD184
Applications Flow Cytometry
Classification Monoclonal
Clone 12G5
Conjugate APC
Description CXCR4; Fusin; SDF-1 receptor; LAP3; LCR1; LESTR; NPYY3R; NPY3R; WHIM; HM89
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen SIVmac variant CP-MAC-infected Sup-T1 cells
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.