missing translation for 'onlineSavingsMsg'
Learn More

CD14 Mouse anti-Human, APC-H7, Clone: MΦP9, BD

Mouse Monoclonal Antibody

£139.36 - £387.23

Specifications

Antigen CD14
Clone MΦP9
Applications Flow Cytometry
Classification Monoclonal
Conjugate APC-H7
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15820069
View Documents
BD Biosciences
560270
25 Tests
ONLINE EXCLUSIVE PRICE
£145.00
£139.36 / Each
Save £5.64
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15889988
View Documents
BD Biosciences
560180
100 Tests
ONLINE EXCLUSIVE PRICE
£404.00
£387.23 / Each
Save £16.77
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Reacts with a 53-55kDa glycosylphosphatidylinositol (GPI)-anchored and single chain glycoprotein expressed at high levels on monocytes. Additionally, CD14 antibody reacts with interfollicular macrophages, reticular dendritic cells and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB. This antibody is suitable for staining acetone-fixed, frozen tissue sections.

Host Species: Mouse
Clone: MφP9
Isotype: IgG2b, κ
Species Reactivity: Human

Flow Cytometry

Specifications

Specifications

CD14
Flow Cytometry
APC-H7
RUO
Human
Human Monocytes
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
MΦP9
Monoclonal
Mouse
Immunology
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
IgG2b κ
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.