Learn More
CD14 Mouse anti-Human, APC-H7, Clone: MΦP9, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 560270
Description
Reacts with a 53-55kDa glycosylphosphatidylinositol (GPI)-anchored and single chain glycoprotein expressed at high levels on monocytes. Additionally, CD14 antibody reacts with interfollicular macrophages, reticular dendritic cells and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB. This antibody is suitable for staining acetone-fixed, frozen tissue sections.
Host Species: Mouse
Clone: MφP9
Isotype: IgG2b, κ
Species Reactivity: Human
Flow Cytometry
Specifications
| CD14 | |
| Monoclonal | |
| APC-H7 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| MΦP9 | |
| Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
| Human Monocytes | |
| 25 Tests | |
| Immunology | |
| Human | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.