Learn More
CD11c Hamster anti-Mouse, APC, Clone: HL3, BD
Hamster Monoclonal Antibody
£107.34 - £251.41
Specifications
| Antigen | CD11c |
|---|---|
| Clone | HL3 |
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15860789
|
BD Biosciences
561119 |
25 μg |
£112.00 £107.34 / Each Save £4.66 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15876198
|
BD Biosciences
550261 |
0.1 mg |
£263.00 £251.41 / Each Save £11.59 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The HL3 antibody reacts with the integrin αx chain of gp150, 95 (CD11c/CD18) which is expressed on dendritic cells and CD4- CD8+ intestinal intraepithelial lymphocytes (IEL) and is upregulated on IEL and lymph-node T cells following in vivo activation. CD11c is also found on human NK cells. Although its expression on mouse NK cells is not published, we have detected CD11c on mouse splenic NK cells. Cells of the monocyte/macrophage lineage have been reported to express low levels of CD11c. CD11c plays a role in binding of iC3b.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Hamster
Clone: HL3
Isotype: IgG1 λ
Species Reactivity: Mouse
Immunogen: C57BL/6 Mouse Intestinal Intraepithelial Lymphocytes
Flow Cytometry
Specifications
| CD11c | |
| 0.2mg/mL | |
| Monoclonal | |
| Hamster | |
| Mouse | |
| C57BL/6 Mouse Intestinal Intraepithelial Lymphocytes | |
| Affinity Purified | |
| Cd11c; Itgax; Integrin alpha-X; Integrin αX; Cr4; Complement receptor 4 |
| HL3 | |
| Flow Cytometry | |
| APC | |
| RUO | |
| Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.