missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11a Rat anti-Mouse, FITC, Clone: 2D7, BD
Rat Monoclonal Antibody
£154.08 - £259.00
Specifications
| Antigen | CD11a |
|---|---|
| Clone | 2D7 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15881229
|
BD Biosciences
561683 |
0.1 mg | |||||||
|
15857008
|
BD Biosciences
553120 |
0.5 mg |
£157.00 £154.08 / Each Save £2.92 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The 2D7 monoclonal antibody specifically binds to the 180-kDa α; L chain of LFA-1 (CD11a/CD18, α; L β; 2 integrin), a heterodimeric surface glycoprotein expressed on almost all leukocytes. CD8a+CD8b- intestinal intraepithelial T lymphocytes, which are believed to be thymus independent, do not express CD11a. LFA-1 mediates a variety of heterotypic and homotypic intracellular adhesions through interaction with ICAM-1 (CD54) and ICAM-2 (CD102), including participation in the immunological synapses between CD8+ T lymphocytes and antigen-presenting cells. mAb 2D7 has been reported to block an in vitro allogeneic mixed-leukocyte reaction. The 2D7 and M17/4 (Cat. No. 553337, for the NA/LE™ format) antibodies are reported to recognize different epitopes of the CD11a molecule.Specifications
| CD11a | |
| 0.5mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| Not Reported | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| 2D7 | |
| Flow Cytometry | |
| FITC | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG2a κ | |
| Affinity Purified | |
| Itgal; Integrin alpha-L; Integrin αL; ITAL; LFA-1A; LFA-1α; Ly-15 |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title