missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11a Rat anti-Mouse, FITC, Clone: 2D7, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 553120
This item is not returnable.
View return policy
Description
The 2D7 monoclonal antibody specifically binds to the 180-kDa α; L chain of LFA-1 (CD11a/CD18, α; L β; 2 integrin), a heterodimeric surface glycoprotein expressed on almost all leukocytes. CD8a+CD8b- intestinal intraepithelial T lymphocytes, which are believed to be thymus independent, do not express CD11a. LFA-1 mediates a variety of heterotypic and homotypic intracellular adhesions through interaction with ICAM-1 (CD54) and ICAM-2 (CD102), including participation in the immunological synapses between CD8+ T lymphocytes and antigen-presenting cells. mAb 2D7 has been reported to block an in vitro allogeneic mixed-leukocyte reaction. The 2D7 and M17/4 (Cat. No. 553337, for the NA/LE™ format) antibodies are reported to recognize different epitopes of the CD11a molecule.
Specifications
| CD11a | |
| Monoclonal | |
| 0.5mg/mL | |
| Itgal; Integrin alpha-L; Integrin αL; ITAL; LFA-1A; LFA-1α; Ly-15 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 2D7 | |
| FITC | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Not Reported | |
| 0.5 mg | |
| Immunology | |
| Mouse | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction