Learn More
CD117 Mouse anti-Human, APC, Clone: YB5.B8, BD
Mouse Monoclonal Antibody
£299.00 - £323.91
Specifications
| Antigen | CD117 |
|---|---|
| Clone | YB5.B8 |
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15850789
|
BD Biosciences
561118 |
25 μg |
£299.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15856268
|
BD Biosciences
550412 |
1 mg |
£338.00 £323.91 / Each Save £14.09 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Recognizes CD117 (the C-Kit), a 145kDa cell-surface glycoprotein with tyrosine kinase activity. CD117 is present on hematopoietic progenitor cell subsets, thymocytes, mast cells, hepatocytes and histiocytes. CD117 serves as a cytokine receptor for steel factor (SLF), also known as stem cell factor (SCF) or mast cell growth factor (MGF). The interaction of c-kit and SLF is crucial to hematopoiesis, mast cell differentiation, melanogenesis, and germ cell development. The ability of YB5.B8 antibody to block the binding of c-kit ligand is still
controversial. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: YB5.B8
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Specifications
| CD117 | |
| 0.2mg/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| Acute myelocytic leukemia blasts | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| YB5.B8 | |
| Flow Cytometry | |
| APC | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified | |
| KIT; c-Kit; SCFR; PBT; Mast/stem cell growth factor receptor |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.