Learn More
CD117 Mouse anti-Human, APC, Clone: YB5.B8, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 561118
Description
Recognizes CD117 (the C-Kit), a 145kDa cell-surface glycoprotein with tyrosine kinase activity. CD117 is present on hematopoietic progenitor cell subsets, thymocytes, mast cells, hepatocytes and histiocytes. CD117 serves as a cytokine receptor for steel factor (SLF), also known as stem cell factor (SCF) or mast cell growth factor (MGF). The interaction of c-kit and SLF is crucial to hematopoiesis, mast cell differentiation, melanogenesis, and germ cell development. The ability of YB5.B8 antibody to block the binding of c-kit ligand is still
controversial. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: YB5.B8
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Specifications
| CD117 | |
| Monoclonal | |
| 0.2mg/mL | |
| KIT; c-Kit; SCFR; PBT; Mast/stem cell growth factor receptor | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| YB5.B8 | |
| APC | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Acute myelocytic leukemia blasts | |
| 25 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.