Learn More
CD107a Mouse anti-Human, PE, Clone: H4A3, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555801
Description
The H4A3 monoclonal antibody specifically binds to the heavily glycosylated 110 kDa Lysosomal-associated membrane protein 1, LAMP-1. LAMP-1 is a widely expressed intracellular antigen. It is also expressed on the surface of activated platelets, PHA-activated lymphocytes, cytotoxic T cells and NK cells, and some tumor cell lines, including U937 and KG1a. LAMP-1 has been shown to be a ligand for E-selectin-mediated cell adhesion. LAMP-1 and LAMP-2 (CD107b) are carriers for poly-N-acetyllactosamines and are able to display sialyl Le[x] termini.
Specifications
| CD107a | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human Adult Adherent Peripheral Blood Cells | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| H4A3 | |
| LAMP1; LAMP-1; LAMPA; LGP120 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.