Learn More
CD107a Mouse anti-Human, PE, Clone: H4A3, BD
Mouse Monoclonal Antibody
£58.70 - £209.04
Specifications
| Antigen | CD107a |
|---|---|
| Clone | H4A3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15820629
|
BD Biosciences
560948 |
25 Tests |
£61.00 £58.70 / Each Save £2.30 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15838448
|
BD Biosciences
555801 |
100 Tests |
£218.00 £209.04 / Each Save £8.96 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The H4A3 monoclonal antibody specifically binds to the heavily glycosylated 110 kDa Lysosomal-associated membrane protein 1, LAMP-1. LAMP-1 is a widely expressed intracellular antigen. It is also expressed on the surface of activated platelets, PHA-activated lymphocytes, cytotoxic T cells and NK cells, and some tumor cell lines, including U937 and KG1a. LAMP-1 has been shown to be a ligand for E-selectin-mediated cell adhesion. LAMP-1 and LAMP-2 (CD107b) are carriers for poly-N-acetyllactosamines and are able to display sialyl Le[x] termini.
Specifications
| CD107a | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified | |
| LAMP1; LAMP-1; LAMPA; LGP120 |
| H4A3 | |
| Monoclonal | |
| Mouse | |
| Human | |
| Human Adult Adherent Peripheral Blood Cells | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.