missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCR2 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 6 publications
£208.00 - £498.00
Specifications
| Antigen | CCR2 |
|---|---|
| Clone | 475301 |
| Dilution | Flow Cytometry 5 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18789708
![]() |
R&D Systems
FAB5538N-025 |
25 Tests |
£208.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18767597
![]() |
R&D Systems
FAB5538N-100 |
100 Tests |
£498.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR2 Monoclonal specifically detects CCR2 in Mouse samples. It is validated for Flow Cytometry.Specifications
| CCR2 | |
| Flow Cytometry 5 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| Mouse | |
| 729230 | |
| L1.2 mouse pro-B cell line transfected with mouse CCR2 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| 475301 | |
| Flow Cytometry | |
| Alexa Fluor 700 | |
| Rat | |
| C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2, CC-CKR-2CKR2B, CCR2, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-R, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor | |
| CCR2 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects mouse and rat CCR2 transfectants but not the parental cell line in flow cytometry. Does not detect HEK293 human embryonic kidney cell line transfected with mouse CCR5 in flow cytometry. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title