Learn More
Mouse CCR2 Alexa Fluor 700-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 6 publications
Brand: R&D Systems FAB5538N-100
This item is not returnable.
View return policy
Description
CCR2 Monoclonal specifically detects CCR2 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CCR2 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2, CC-CKR-2CKR2B, CCR2, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-R, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects mouse and rat CCR2 transfectants but not the parental cell line in flow cytometry. Does not detect HEK293 human embryonic kidney cell line transfected with mouse CCR5 in flow cytometry. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 475301 | |
| Flow Cytometry 5 uL/10^6 cells | |
| CCR2 | |
| L1.2 mouse pro-B cell line transfected with mouse CCR2 | |
| 100 Tests | |
| 729230 | |
| Mouse | |
| Purified |