missing translation for 'onlineSavingsMsg'
Learn More

Caspase-8 Mouse anti-Human, Unlabeled, Clone: 3-1-9, BD

Mouse Monoclonal Antibody

£178.90 - £404.00

Specifications

Antigen Caspase-8
Clone 3/1/2009
Concentration 0.5mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15816588
View Documents
BD Biosciences
551242
50 μg
ONLINE EXCLUSIVE PRICE
£187.00
£178.90 / Each
Save £8.10
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15826588
View Documents
BD Biosciences
551243
150 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Caspase-8 (FLICE/MACH-1) is a 55kDa cytosolic protein with homology to the CD95/Fas-associated signal transducer, FADD/MORT-1, as well as to other caspase (ICE/Ced-3) cysteine proteases. The N-terminal region of caspase-8 contains an amino acid sequence, termed the death domain, that facilitates caspase-8-FADD direct interaction. FADD therefore acts as an adapter molecule, allowing caspase-8 to become recruited to the cytoplasmic region of Fas following receptor activation. Viral proteins (v-FLIPS) which inhibit recruitment and activation of
caspase-8 have been isolated. Caspase-8 is produced as a proenzyme (55/50kDa doublet) which upon receptor aggregation is proteolytically cleaved into smaller subunits of 40/36 (doublet), and 23kDa. Overexpression of caspase-8 is sufficient to induce apoptosis in certain cell lines (e.g., MCF-7) and this phenotype is blocked by overexpression of the caspase-3 protease inhibitor, CrmA. The antibody recognizes both the proform of human caspase-8 (55/50kDa doublet) as well as the cleaved forms which migrate at 40/36 (doublet) and 23kDa in SDS/PAGE. It
also immunoprecipitates the full-length human caspase-8. Full-length recombinant human caspase-8 protein was used as immunogen.

Host Species: Mouse
Clone: 3-1-9
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Human Caspase-8 full-length recombinant protein

Immunoprecipitation, Western Blotting

Specifications

Specifications

Caspase-8
0.5mg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing ≤0.09% sodium azide.
IgG1 κ
Affinity Purified
FLICE, MACH-1, Mch5
3/1/2009
Western Blot
Unconjugated
RUO
Human
Human Caspase-8 full-length recombinant protein
Primary
Store undiluted at 4°C
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.