missing translation for 'onlineSavingsMsg'
Learn More

Caspase-8 Mouse anti-Human, Unlabeled, Clone: 3-1-9, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 551242

 View more versions of this product

Product Code. 15816588

  • ONLINE EXCLUSIVE PRICE
    £187.00
    £178.90 / Each
    Save £8.10
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Caspase-8 (FLICE/MACH-1) is a 55kDa cytosolic protein with homology to the CD95/Fas-associated signal transducer, FADD/MORT-1, as well as to other caspase (ICE/Ced-3) cysteine proteases. The N-terminal region of caspase-8 contains an amino acid sequence, termed the death domain, that facilitates caspase-8-FADD direct interaction. FADD therefore acts as an adapter molecule, allowing caspase-8 to become recruited to the cytoplasmic region of Fas following receptor activation. Viral proteins (v-FLIPS) which inhibit recruitment and activation of
caspase-8 have been isolated. Caspase-8 is produced as a proenzyme (55/50kDa doublet) which upon receptor aggregation is proteolytically cleaved into smaller subunits of 40/36 (doublet), and 23kDa. Overexpression of caspase-8 is sufficient to induce apoptosis in certain cell lines (e.g., MCF-7) and this phenotype is blocked by overexpression of the caspase-3 protease inhibitor, CrmA. The antibody recognizes both the proform of human caspase-8 (55/50kDa doublet) as well as the cleaved forms which migrate at 40/36 (doublet) and 23kDa in SDS/PAGE. It
also immunoprecipitates the full-length human caspase-8. Full-length recombinant human caspase-8 protein was used as immunogen.

Host Species: Mouse
Clone: 3-1-9
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Human Caspase-8 full-length recombinant protein

Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Caspase-8
Monoclonal
0.5mg/mL
FLICE, MACH-1, Mch5
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C
Western Blot
3/1/2009
Unconjugated
Aqueous buffered solution containing ≤0.09% sodium azide.
Human Caspase-8 full-length recombinant protein
50 μg
Cell Biology
Human
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.