Learn More
Caspase-2 Mouse anti-Human, Unlabeled, Clone: 35, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 611023
Description
Caspase-2/ICH-1 is related to the C. elegans cell death gene product CED-3 and its mammalian homologue interleukin-1β-converting enzyme (ICE). Caspase-2 /ICH-1 was identified from a mouse cDNA library and originally termed NEDD-2. The NEDD-2 mRNA was found to be expressed during early mouse embryonic brain development and subsequently down-regulated in adult neuronal tissue. With the identification of the human NEDD-2 gene, the murine gene was renamed Ich-1 to symbolize Ice and ced-3 homology. Caspase-2/ICH-1 mRNA is alternatively spliced. The larger mRNA species encoding a product of 435 amino acids is known as Caspase-2 long, or ICH-1L. The smaller mRNA species encoding a protein of 312 amino acids is named Caspase-2 short, or ICH-1S. Overexpression of ICH-1L induces apoptosis, while over-expression of Ich-1S suppresses Rat-1 cell death induced by serum deprivation. Thus, it appears that Caspase-2/ICH-1 plays an important dual role in programmed cell death.
Immunofluorescence, Western Blotting
Specifications
| Caspase-2 | |
| Monoclonal | |
| 250μg/mL | |
| ICH-1L | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
| Western Blot | |
| 35 | |
| Unconjugated | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Human ICH-1L aa. 225-401 | |
| 150 μg | |
| Cell Biology | |
| Human | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.