missing translation for 'onlineSavingsMsg'
Learn More

Caspase-2 Mouse anti-Human, Unlabeled, Clone: 35, BD

Product Code. p-3851831
Change view
Click to view available options
Quantity:
150 μg
50 μg
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15835769 50 μg 1 set
15845769 150 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15835769 Supplier BD Biosciences Supplier No. 611022

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Caspase-2/ICH-1 is related to the C. elegans cell death gene product CED-3 and its mammalian homologue interleukin-1β-converting enzyme (ICE). Caspase-2 /ICH-1 was identified from a mouse cDNA library and originally termed NEDD-2. The NEDD-2 mRNA was found to be expressed during early mouse embryonic brain development and subsequently down-regulated in adult neuronal tissue. With the identification of the human NEDD-2 gene, the murine gene was renamed Ich-1 to symbolize Ice and ced-3 homology. Caspase-2/ICH-1 mRNA is alternatively spliced. The larger mRNA species encoding a product of 435 amino acids is known as Caspase-2 long, or ICH-1L. The smaller mRNA species encoding a protein of 312 amino acids is named Caspase-2 short, or ICH-1S. Overexpression of ICH-1L induces apoptosis, while over-expression of Ich-1S suppresses Rat-1 cell death induced by serum deprivation. Thus, it appears that Caspase-2/ICH-1 plays an important dual role in programmed cell death.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Caspase-2
Applications Western Blot
Classification Monoclonal
Clone 35
Concentration 250μg/mL
Conjugate Unconjugated
Description ICH-1L
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human ICH-1L aa. 225-401
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.