missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Defensin 3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 16 publications
£145.00 - £421.00
Specifications
| Antigen | beta-Defensin 3 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18765983
|
Novus Biologicals
NB200-117SS |
0.025 mL |
£145.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18701514
|
Novus Biologicals
NB200-117 |
£421.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
beta-Defensin 3 Polyclonal specifically detects beta-Defensin 3 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).Specifications
| beta-Defensin 3 | |
| Western Blot 1:500-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Polyclonal | |
| Rabbit | |
| Immunology | |
| PBS with 0.02% Sodium Azide | |
| BD3, BD-3, beta-defensin 103, Beta-defensin 3, DEFB103, DEFB-3, DEFB3DEFB103A, Defensin, beta 103, defensin, beta 103B, Defensin-like protein, hBD-3, HBD3HBP3 | |
| DEFB103B | |
| IgG | |
| Affinity Purified | |
| Human and rat (PMID: 21369366). |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Rat | |
| P81534 | |
| 55894 | |
| A synthetic peptide made to an internal portion of Defensin beta 3 (between amino acids 10-50) [UniProt P81534] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 7.7 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title