missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Defensin 3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 16 publications
Brand: Novus Biologicals NB200-117SS
This item is not returnable.
View return policy
Description
beta-Defensin 3 Polyclonal specifically detects beta-Defensin 3 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications
| beta-Defensin 3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| BD3, BD-3, beta-defensin 103, Beta-defensin 3, DEFB103, DEFB-3, DEFB3DEFB103A, Defensin, beta 103, defensin, beta 103B, Defensin-like protein, hBD-3, HBD3HBP3 | |
| Rabbit | |
| 7.7 kDa | |
| 0.025 mL | |
| Immunology | |
| 55894 | |
| Human, Rat | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1:500-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| P81534 | |
| DEFB103B | |
| A synthetic peptide made to an internal portion of Defensin beta 3 (between amino acids 10-50) [UniProt P81534] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human and rat (PMID: 21369366). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction