missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-X set Mouse, Unlabeled, Clone: 2H12, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 551020
This item is not returnable.
View return policy
Description
Members of the Bcl-2 family play a major role in regulating the response of cells to a wide variety of apoptotic signals. Some Bcl-2 family members like Bcl-2, block apoptosis, whereas others promote apoptosis and inhibit Bcl-2 activity. Bcl-x is a Bcl-2 homologue that has two isoforms, resulting from alternative splicing. Bcl-xL (long) is a 241 amino acid protein (25-29 kDa) that is 47% homologous to Bcl-2 on the amino acid level. Bcl-xS (short) is a 178 amino acid protein (∼ 19.5 kDa) lacking a 63 amino acid domain that is well conserved among members of the Bcl-2 protein family. Bcl-xL blocks cell death, whereas Bcl-xS inhibits Bcl-2 and promotes cell death.
Specifications
| Bcl-X set | |
| Monoclonal | |
| Unconjugated | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
| Western Blot | |
| 2H12 | |
| Mouse | |
| 50 μg | |
| Cell Biology | |
| Human, Mouse | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction