missing translation for 'onlineSavingsMsg'
Learn More

Annexin VI Mouse anti-Human, Mouse, Rat, Rabbit, Pig, Clone: 2H7G7, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen Annexin VI
Clone 2H7G7
Concentration 500 μg/mL
Applications Western Blot, Immunofluorescence, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17689766
View Documents
Proteintech
68086-1-IG-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17679766
View Documents
Proteintech
68086-1-IG-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Annexin VI Monoclonal antibody apecifically detetects Annexin VI antigen in Human, Mouse, Rat, Rabbit, Pig Samples. It is validated for Western Blot, Immunofluorescence, ELISA.
Specifications

Specifications

Annexin VI
500 μg/mL
Monoclonal
Liquid
Human, Mouse, Rat, Rabbit, Pig
BC017046
ANXA6
IgG1
Protein G purification
2H7G7
Western Blot, Immunofluorescence, ELISA
Unconjugated
Mouse
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
309
Annexin VI fusion protein Ag3263
Primary
Store at -20°C. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.