missing translation for 'onlineSavingsMsg'
Learn More

Annexin VI Mouse anti-Human, Mouse, Rat, Rabbit, Pig, Clone: 2H7G7, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 68086-1-IG-150UL

 View more versions of this product

Product Code. 17679766

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Annexin VI Monoclonal antibody apecifically detetects Annexin VI antigen in Human, Mouse, Rat, Rabbit, Pig Samples. It is validated for Western Blot, Immunofluorescence, ELISA.
Specifications

Specifications

Annexin VI
Monoclonal
500 μg/mL
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
ANXA6
Annexin VI fusion protein Ag3263
150 μL
309
Store at -20°C. Stable for one year after shipment.
IgG1
Western Blot, Immunofluorescence, ELISA
2H7G7
Unconjugated
BC017046
Mouse
Protein G purification
Primary
Human, Mouse, Rat, Rabbit, Pig
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.