missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Annexin V-FITC Apoptosis Detection Kit, for in vitro diagnostic use
Click to view available options
Quantity:
300 Tests
Unit Size:
300 tests
Description
Includes
- Annexin V-FITC protein
- Binding Buffer (4x)
- Propidium Iodide (20 ug/mL)
- Blue Laser
- Excitation at 488nm
- Emission at 520nm
Specifications
Specifications
| Quantity | 300 Tests |
| Assay | >98% |
| Formula Weight | 35.8 kd |
| Synonym | Lipocortin V, Endonexin II, Calphobindin I |
| For Use With (Application) | Flow Cytometry |
| Conjugate | FITC |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction