missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Annexin V-FITC
Description
Identifies apoptosis at an earlier stage than assays based on nuclear changes such as DNA fragmentation.
- Format: FITC
Flow Cytometry
Specifications
Specifications
| Quantity | 200 Tests |
| Sample Volume | 5 μL |
| Storage Buffer | Aqueous buffered solution containing BSA and ≤0.09% sodium azide |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Conjugate | FITC |
| Form | Liquid |
Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?