missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Annexin V Conjugates for Apoptosis Detection

Product Code. 10443442
Click to view available options
Excitation/Emission:
346/442
410/455
494/518
495/519
555/565
565/578
578/603
590/617
650/660
650/665
679/702
Flow Cytometer Laser Lines:
405
488
488, 532, 561
532
532, 561
633-637
UV
Conjugate:
APC (Allophycocyanin)
Alexa Fluor 350
Alexa Fluor 488
Alexa Fluor 555
Alexa Fluor 568
Alexa Fluor 594
Alexa Fluor 647
Alexa Fluor 680
Biotin-X
FITC
PE
Pacific Blue
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10443442

Brand: Molecular Probes™ A13203

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Detect early stages of apoptosis with Annexin V stand-alone Alexa Fluor, APC, Pacific Blue, PE, FITC, and biotin conjugates using flow cytometry.

Achieve quick and reliable detection of early cell apoptosis with Annexin V stand-alone conjugates for apoptosis detection. Annexin V conjugates offer up to 100-fold difference in fluorescence signal intensity between apoptotic and non-apoptotic cells using flow cytometry.

Annexin V has a high affinity for phosphatidylserine (PS), which becomes exposed on the outer leaflet of cells undergoing apoptosis. Because of this affinity, fluorescently labeled annexin V reagents are commonly used in apoptosis research.

Annexin V conjugates provide quick and reliable detection methods for studying the externalization of phosphatidylserine, an indicator of intermediate stages of apoptosis. The difference in fluorescence intensity between apoptotic and nonapoptotic cells stained with our fluorescent annexin V conjugates, as measured by flow cytometry, is typically about 100-fold.

In collaboration with Nexins Research BV, we provide the best and brightest annexin V conjugates available, including Alexa Fluor 350, 488, 555, 568, 594, 647, and 680 annexin V conjugates, as well as Annexin V APC, Biotin-X, FITC, Pacific Blue, and PE conjugates. Highly fluorescent annexin V conjugates provide quick and reliable detection methods for studying the externalization of phosphatidylserine, one of the earliest indicators of apoptosis.

The Annexin V Pacific Blue conjugate is violet excitable, making it ideal for instruments with a violet laser and for multicolor experiments that include green- or red-fluorescent dyes.

The benefits of our annexin V conjugates include:
• Conjugated to Invitrogen Alexa Fluor and eFluor dyes for brighter signals
• Conjugates for all available lasers
• Available as stand-alone reagents or easy-to-use kits

Annexin V staining to detect apoptotic cells can only be done on live cells and tissue. If samples are to be fixed post-staining, there are specific conditions required to achieve transient retention of signal. These include use of an alcohol-free, aldehyde-based fixation method, use of buffers containing Ca2+ and avoidance of surfactants/detergents. For your convenience, we also offer a concentrated annexin-binding buffer that facilitates the binding of annexin V to phosphatidylserine in apoptosis assays.

TRUSTED_SUSTAINABILITY

Specifications

Description Annexin V, Alexa Fluor 594 conjugate
Quantity 500 μL
Kit Contents Contains 1 vial of annexin V, Alexa Fluor 594 conjugate.
Product Type Annexin V conjugate
Content And Storage Store in refrigerator (2°C to 8°C) and protect from light.
Flow Cytometer Laser Lines 532
Excitation/Emission 590/617
No. of Reactions 100
Color Red
Shipping Condition Wet Ice
Conjugate Alexa Fluor 594
For Use With (Equipment) Flow Cytometer
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.