missing translation for 'onlineSavingsMsg'
Learn More

Annexin A4 Rabbit anti-Human, Mouse, Rat, Clone: 8T6E6, Novus Biologicals™

Rabbit Monoclonal Antibody

£150.00 - £386.00

Specifications

Antigen Annexin A4
Clone 8T6E6
Dilution Western Blot, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Applications Western Blot, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18310484
View Documents
Novus Biologicals
NBP3-16785-20UL
20 μg
£150.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18334264
View Documents
Novus Biologicals
NBP3-16785-100UL
100 μg
£386.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Annexin A4 Monoclonal antibody specifically detects Annexin A4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

Annexin A4
Western Blot, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Monoclonal
Purified
RUO
Human, Mouse, Rat
annexin A4, Annexin IV, annexin IV (placental anticoagulant protein II), annexin-4, ANX4Carbohydrate-binding protein p33/p41, Chromobindin-4, chromobindin-4,35-beta calcimedin, DKFZp686H02120, Endonexin I, Lipocortin IV, MGC75105, P32.5, PAP-II, PIG28, Placental anticoagulant protein II, PP4-X, proliferation-inducing gene 28, proliferation-inducing protein 28, Protein II, ZAP36
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Annexin A4/Annexin A4 (P09525).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
8T6E6
Western Blot, Immunofluorescence
Unconjugated
Rabbit
Cancer
PBS, 0.05% BSA, 50% glycerol, pH7.3
307
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.