missing translation for 'onlineSavingsMsg'
Learn More

Annexin A4 Rabbit anti-Human, Mouse, Rat, Clone: 8T6E6, Novus Biologicals™

Product Code. 18310484 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18310484 20 μg 20µL
18334264 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18310484 Supplier Novus Biologicals Supplier No. NBP31678520UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Annexin A4 Monoclonal antibody specifically detects Annexin A4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Annexin A4
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 8T6E6
Conjugate Unconjugated
Dilution Western Blot, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias annexin A4, Annexin IV, annexin IV (placental anticoagulant protein II), annexin-4, ANX4Carbohydrate-binding protein p33/p41, Chromobindin-4, chromobindin-4,35-beta calcimedin, DKFZp686H02120, Endonexin I, Lipocortin IV, MGC75105, P32.5, PAP-II, PIG28, Placental anticoagulant protein II, PP4-X, proliferation-inducing gene 28, proliferation-inducing protein 28, Protein II, ZAP36
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Annexin A4/Annexin A4 (P09525).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 307
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.