missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Annexin A10 Recombinant Rabbit Monoclonal Antibody (23GB3215), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | Annexin A10 |
|---|---|
| Clone | 23GB3215 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088265
![]() |
Invitrogen™
MA553606 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088291
|
Invitrogen™
MA553607 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Annexin A10 Recombinant Monoclonal antibody specifically detects Annexin A10 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| Annexin A10 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q9QZ10, Q9UJ72 | |
| 11199, 26359, 498622 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3215 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| annexin 14; annexin A10; Annexin-10; annexin-14; ANX14; Anxa10; RGD1560956 | |
| A synthetic peptide of human Annexin A10 (50-200AA). | |
| Primary | |
| -20°C | |
| ANXA10 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title