missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Annexin A10 Recombinant Rabbit Monoclonal Antibody (23GB3215), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553607
This item is not returnable.
View return policy
Description
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. The function of this gene has not yet been determined.
Specifications
| Annexin A10 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ANXA10 | |
| annexin 14; annexin A10; Annexin-10; annexin-14; ANX14; Anxa10; RGD1560956 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11199, 26359, 498622 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3215 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9QZ10, Q9UJ72 | |
| ANXA10 | |
| A synthetic peptide of human Annexin A10 (50-200AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction