missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Annexin A10 Monoclonal Antibody (K94033_8E3), Invitrogen™

Mouse Monoclonal Antibody

£380.00 - £570.00

Specifications

Antigen Annexin A10
Clone K94033_8E3
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30204360
View Documents
Invitrogen™
MA555701
50 μL
£380.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30210524
View Documents
Invitrogen™
MA556081
200 μL
£570.00
200µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sequence of this protein is as follows: SMYGRDLIGD MREQLSDHFK DVMAGLMYPP PLYDAHELWH AMKGVGTDEN CLIEILASRT NGEIFQMREA YCLQYSNNLQ EDIYSETSGH FRDTLMNLVQ GTREEGYTDP AMAAQDAMVL WEACQQKTGE HKTMLQMILC NKSYQQLRLV FQEFQNISGQ DMVDAINECY DGYFQELLVA IVLCVRDKPA YFAYRLYSAI HDFGFHNKTV IRILIARSEI DLLTIRKRYK ERYGKSLFHD IRNFASGHYK

Annexin A10 Monoclonal antibody specifically detects Annexin A10 in Human samples. It is validated for Western Blot
Specifications

Specifications

Annexin A10
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
annexin 14; annexin A10; Annexin-10; annexin-14; ANX14; Anxa10; RGD1560956
Recombinant human ANXA10 (immunogen range is 61-310).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
ANXA10
K94033_8E3
Western Blot
Unconjugated
Mouse
Human
Q9UJ72
11199
IgG1
Protein A
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.