missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Annexin A10 Monoclonal Antibody (K94033_8E3), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA556081

 View more versions of this product

Product Code. 30210524

  • £570.00 / 200µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence of this protein is as follows: SMYGRDLIGD MREQLSDHFK DVMAGLMYPP PLYDAHELWH AMKGVGTDEN CLIEILASRT NGEIFQMREA YCLQYSNNLQ EDIYSETSGH FRDTLMNLVQ GTREEGYTDP AMAAQDAMVL WEACQQKTGE HKTMLQMILC NKSYQQLRLV FQEFQNISGQ DMVDAINECY DGYFQELLVA IVLCVRDKPA YFAYRLYSAI HDFGFHNKTV IRILIARSEI DLLTIRKRYK ERYGKSLFHD IRNFASGHYK

This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. The function of this gene has not yet been determined.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Annexin A10
Monoclonal
1 mg/mL
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Q9UJ72
ANXA10
Recombinant human ANXA10 (immunogen range is 61-310).
200 μL
Primary
Human
Antibody
IgG1
Western Blot
K94033_8E3
Unconjugated
ANXA10
annexin 14; annexin A10; Annexin-10; annexin-14; ANX14; Anxa10; RGD1560956
Mouse
Protein A
RUO
11199
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.