missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANLN Mouse anti-Human, Clone: 1B4A8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66643-1-IG-20UL
This item is not returnable.
View return policy
Description
ZFP281, also known as ZBP-99, is a member of a conserved family of transcription factors that interact with GC-rich promoters. It contains four Kruppel-like zinc fingers and is highly homologous to ZBP-89, another regulator of ornithine decarboxylase gene expression. ZFP281 also interacts with many other other transcription factors, such as Sp1 to repress vimentin gene transcription. ZFP281 has also been shown to control embryonic stem cell pluripotency by direct regulating specific target genes. ZFP281 phyically interacts with the transcription factos POU5F1, Sox2, and NANOG. Gene expression microarray anlysis indicated that some ZFP281 target genes were activated, while others were repressed upon knockdown of ZFP281 expression, showing that ZFP281 plays bifuctinal roles in regulating gene expression in stem cells.Specifications
| ANLN | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9NQW6 | |
| ANLN | |
| ANLN Fusion Protein Ag19294 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 1B4A8 | |
| Unconjugated | |
| ANLN | |
| Actin binding protein anillin, anillin, actin binding protein, ANLN, scra, Scraps | |
| Mouse | |
| Protein G | |
| RUO | |
| 54443 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction