missing translation for 'onlineSavingsMsg'
Learn More

ANLN Mouse anti-Human, Clone: 1B4A8, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66643-1-IG-150UL

 View more versions of this product

Product Code. 16818715

  • £359.47 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ZFP281, also known as ZBP-99, is a member of a conserved family of transcription factors that interact with GC-rich promoters. It contains four Kruppel-like zinc fingers and is highly homologous to ZBP-89, another regulator of ornithine decarboxylase gene expression. ZFP281 also interacts with many other other transcription factors, such as Sp1 to repress vimentin gene transcription. ZFP281 has also been shown to control embryonic stem cell pluripotency by direct regulating specific target genes. ZFP281 phyically interacts with the transcription factos POU5F1, Sox2, and NANOG. Gene expression microarray anlysis indicated that some ZFP281 target genes were activated, while others were repressed upon knockdown of ZFP281 expression, showing that ZFP281 plays bifuctinal roles in regulating gene expression in stem cells.
Specifications

Specifications

ANLN
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9NQW6
ANLN
ANLN Fusion Protein Ag19294
150 μL
Primary
Human
Antibody
IgG1
Western Blot
1B4A8
Unconjugated
ANLN
Actin binding protein anillin, anillin, actin binding protein, ANLN, scra, Scraps
Mouse
Protein G
RUO
54443
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.