missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ankyrin Brain Antibody (S105-13), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59374-25ug
This item is not returnable.
View return policy
Description
Ankyrin Brain Monoclonal specifically detects Ankyrin Brain in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Ankyrin Brain | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 50% glycerol with 0.09% Sodium Azide | |
| ANK2 | |
| Synthetic peptide amino acids 203-496 of human Ankyrin-B | |
| 25 μg | |
| Primary | |
| Detects 200kDa. No cross reactivity against Ankyrin-G. | |
| Store at -20°C. | |
| IgG1 |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| S105-13 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Ank2, Ankyrin 2, Ankyrin 2 brain, Ankyrin B, Ankyrin brain, Ankyrin nonerythroid, Brain Ankyrin | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 287 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction