missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ankyrin Brain Antibody (S105-13), Novus Biologicals™
Mouse Monoclonal Antibody
£168.00 - £427.00
Specifications
| Antigen | Ankyrin Brain |
|---|---|
| Clone | S105-13 |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18255325
|
Novus Biologicals
NBP2-59374 |
100 μg |
£427.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18669027
|
Novus Biologicals
NBP2-59374-25ug |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ankyrin Brain Monoclonal specifically detects Ankyrin Brain in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| Ankyrin Brain | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| 287 | |
| Synthetic peptide amino acids 203-496 of human Ankyrin-B | |
| Primary | |
| Detects 200kDa. No cross reactivity against Ankyrin-G. |
| S105-13 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Ank2, Ankyrin 2, Ankyrin 2 brain, Ankyrin B, Ankyrin brain, Ankyrin nonerythroid, Brain Ankyrin | |
| ANK2 | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts