Learn More
Invitrogen™ Ankyrin B Monoclonal Antibody (K52014_1E12), Invitrogen™
Mouse Monoclonal Antibody
£416.00 - £734.00
Specifications
| Antigen | Ankyrin B |
|---|---|
| Clone | K52014_1E12 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30212853
|
Invitrogen™
MA555908 |
50 μL |
£416.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30206488
|
Invitrogen™
MA556288 |
200 μL |
£734.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: DSGEKFNGSS QRRKRPKKSD SNASFLRAAR AGNLDKVVEY LKGGIDINTC NQNGLNALHL AAKEGHVGLV QELLGRGSSV DSATKKGNTA LHIASLAGQA EVVKVLVKEG ANINAQSQNG FTPLYMAAQE NHIDVVKYLL ENGANQSTAT EDGFTPLAVA LQQGHNQAVA ILLENDTKGK VRLPALHIAA RKDDTKSAAL LLQNDHNADV QSKMMVNRTT ESGFTPLHIA AHYGNVNVAT LLLNRGAAVD FTARNGITPL HVASKRGNTN MVKLLLDRGG QIDAKTRDGL TPLHCAARSG
Ankyrin B Monoclonal antibody specifically detects Ankyrin B in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| Ankyrin B | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| AI835472; Ank2; ANK-2; Ank3; AnkB; ankyrin 2, brain; ankyrin 2, neuronal; ankyrin 3, epithelial; ankyrin B; ankyrin, brain; ankyrin-2; ankyrin-2, nonerythrocytic; ankyrin-B; AW491075; Brain ankyrin; brank-2; LQT4; Non-erythroid ankyrin | |
| Recombinant human ANK2 (immunogen range is 11-310). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| ANK2 |
| K52014_1E12 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q01484 | |
| 287 | |
| IgG2a | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.