missing translation for 'onlineSavingsMsg'
Learn More

ANKRD13C Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen ANKRD13C
Concentration 0.17 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16822605
View Documents
Proteintech
24000-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16812605
View Documents
Proteintech
24000-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Acts as a molecular chaperone for G protein-coupled receptors, regulating their biogenesis and exit from the ER.
Specifications

Specifications

ANKRD13C
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Mouse, Human
Q3UX43, Q8N6S4
433667, 81573
ANKRD13C Fusion Protein Ag21202
Primary
-20°C
ANKRD13C
0.17 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
ANKRD13C, ankyrin repeat domain 13C, RP4 677H15.5
Ankrd13c
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.