missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD13C Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24000-1-AP-150UL
This item is not returnable.
View return policy
Description
Acts as a molecular chaperone for G protein-coupled receptors, regulating their biogenesis and exit from the ER.Specifications
| ANKRD13C | |
| Polyclonal | |
| Unconjugated | |
| ANKRD13C | |
| ANKRD13C, ankyrin repeat domain 13C, RP4 677H15.5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 433667, 81573 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3UX43, Q8N6S4 | |
| Ankrd13c | |
| ANKRD13C Fusion Protein Ag21202 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction