missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKMY2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-55412
This item is not returnable.
View return policy
Description
ANKMY2 Polyclonal specifically detects ANKMY2 in Human samples. It is validated for Western Blot.
Specifications
| ANKMY2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| ankyrin repeat and MYND domain containing 2, ankyrin repeat and MYND domain-containing protein 2, DKFZP564O043, ZMYND20 | |
| Rabbit | |
| 49 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge vial prior to reconstitution. Add 50μL distilled water to a final antibody concentration of 1mg/mL. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q8IV38 | |
| ANKMY2 | |
| Synthetic peptides corresponding to ANKMY2(ankyrin repeat and MYND domain containing 2) Antibody(against the N terminal of ANKMY2 (NP_064715). Peptide sequence MVHIKKGELTQEEKELLEVIGKGTVQEAGTLLSSKNVRVNCLDENGMTPL. | |
| Affinity purified | |
| RUO | |
| 57037 | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Yeast | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction