Learn More
Anhydrotetracycline hydrochloride, Thermo Scientific™
Anhydrotetracycline hydrochloride, CAS # 13803-65-1, is a tetracycline derivative showing antibacterial activity commonly used to control tetracycline-dependent gene expression systems in bacteria.
£50.65 - £94.65
Chemical Identifiers
| CAS | 13803-65-1 |
|---|---|
| Molecular Formula | C22H23ClN2O6 |
| Molecular Weight (g/mol) | 446.88 |
| MDL Number | MFCD00151453 |
| InChI Key | FFZXKJVSZDKEMY-XGRJIHFXNA-N |
| PubChem CID | 54710409 |
| IUPAC Name | hydrogen (2Z)-2-[amino(hydroxy)methylidene]-4-(dimethylamino)-11,12a-dihydroxy-6-methyl-1,2,3,4,4a,5,12,12a-octahydrotetracene-1,3,12-trione chloride |
| SMILES | [H+].[Cl-].CN(C)C1C2CC3=C(C(O)=C4C=CC=CC4=C3C)C(=O)C2(O)C(=O)\C(=C(\N)O)C1=O |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15468139
|
Thermo Scientific Alfa Aesar
J66688.MA |
10mg |
£55.90 £50.65 / 10mg Save £5.25 9% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15478139
|
Thermo Scientific Alfa Aesar
J66688.MB |
25mg |
£105.00 £94.65 / 25mg Save £10.35 9% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
• Anhydrotetracycline hydrochloride is a degradation product derived from tetracycline hydrochloride. It acts as an effector of tetracycline-dependent gene expression in bacterial expression systems. In this system the transcription of genes is reversibly turned on/off in the presence of several compounds including, anhydrotetracycline hydrochloride.
• This compound poorly binds to the 30S ribosomal subunit compared with its precursor, tetracycline.
Applications
• Anhydrotetracycline hydrochloride is commonly used in all fields of biology for control of transcription or translation processes
• This compound has been used in laboratory experiments and shows lower antibacterial activity against Escherichia coli compared with tetracycline.
Chemical Identifiers
| 13803-65-1 | |
| 446.88 | |
| FFZXKJVSZDKEMY-XGRJIHFXNA-N | |
| hydrogen (2Z)-2-[amino(hydroxy)methylidene]-4-(dimethylamino)-11,12a-dihydroxy-6-methyl-1,2,3,4,4a,5,12,12a-octahydrotetracene-1,3,12-trione chloride |
| C22H23ClN2O6 | |
| MFCD00151453 | |
| 54710409 | |
| [H+].[Cl-].CN(C)C1C2CC3=C(C(O)=C4C=CC=CC4=C3C)C(=O)C2(O)C(=O)\C(=C(\N)O)C1=O |
Specifications
| 13803-65-1 | |
| Moisture sensitive | |
| MFCD00151453 | |
| Soluble in ethanol, DMSO, or DMF | |
| [H+].[Cl-].CN(C)C1C2CC3=C(C(O)=C4C=CC=CC4=C3C)C(=O)C2(O)C(=O)\C(=C(\N)O)C1=O | |
| 446.88 | |
| 462.88 |
| Yellow | |
| C22H23ClN2O6 | |
| 4228856 | |
| FFZXKJVSZDKEMY-XGRJIHFXNA-N | |
| hydrogen (2Z)-2-[amino(hydroxy)methylidene]-4-(dimethylamino)-11,12a-dihydroxy-6-methyl-1,2,3,4,4a,5,12,12a-octahydrotetracene-1,3,12-trione chloride | |
| 54710409 | |
| Anhydrotetracycline hydrochloride |
Safety and Handling
P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
H319-H361d
missing translation for 'rtecsNumber' : QI7860000
missing translation for 'tsca' : No
missing translation for 'storageNote' : -30° to -10°C
Recommended Storage : Store at -20°C; Store under Argon
- Anhydrotetracycline hydrochloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H361d-Suspected of damaging the unborn child.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contain: Anhydrotetracyclinehydrochloride
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.