missing translation for 'onlineSavingsMsg'
Learn More

Anhydrotetracycline hydrochloride, Thermo Scientific™

Anhydrotetracycline hydrochloride, CAS # 13803-65-1, is a tetracycline derivative showing antibacterial activity commonly used to control tetracycline-dependent gene expression systems in bacteria.

Brand:  Thermo Scientific Alfa Aesar J66688.MA


Quantity 10mg

 View more versions of this product

Product Code. 15468139

  • ONLINE EXCLUSIVE PRICE
    £55.90
    £50.65 / 10mg
    Save £5.25
    9% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description

• Anhydrotetracycline hydrochloride is a degradation product derived from tetracycline hydrochloride. It acts as an effector of tetracycline-dependent gene expression in bacterial expression systems. In this system the transcription of genes is reversibly turned on/off in the presence of several compounds including, anhydrotetracycline hydrochloride.
• This compound poorly binds to the 30S ribosomal subunit compared with its precursor, tetracycline.

Applications
• Anhydrotetracycline hydrochloride is commonly used in all fields of biology for control of transcription or translation processes
• This compound has been used in laboratory experiments and shows lower antibacterial activity against Escherichia coli compared with tetracycline.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

13803-65-1
446.88
FFZXKJVSZDKEMY-XGRJIHFXNA-N
hydrogen (2Z)-2-[amino(hydroxy)methylidene]-4-(dimethylamino)-11,12a-dihydroxy-6-methyl-1,2,3,4,4a,5,12,12a-octahydrotetracene-1,3,12-trione chloride
C22H23ClN2O6
MFCD00151453
54710409
[H+].[Cl-].CN(C)C1C2CC3=C(C(O)=C4C=CC=CC4=C3C)C(=O)C2(O)C(=O)\C(=C(\N)O)C1=O

Specifications

13803-65-1
10mg
C22H23ClN2O6
4228856
FFZXKJVSZDKEMY-XGRJIHFXNA-N
hydrogen (2Z)-2-[amino(hydroxy)methylidene]-4-(dimethylamino)-11,12a-dihydroxy-6-methyl-1,2,3,4,4a,5,12,12a-octahydrotetracene-1,3,12-trione chloride
54710409
Anhydrotetracycline hydrochloride
Yellow
Moisture sensitive
MFCD00151453
Soluble in ethanol, DMSO, or DMF
[H+].[Cl-].CN(C)C1C2CC3=C(C(O)=C4C=CC=CC4=C3C)C(=O)C2(O)C(=O)\C(=C(\N)O)C1=O
446.88
462.88
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Anhydrotetracycline hydrochloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H361d-Suspected of damaging the unborn child.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contain: Anhydrotetracyclinehydrochloride

SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.