Learn More
Sampling Systems™ Angular Beaker
Product Code. 17992032 Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
250 mL
500 mL
1 L
2 L
Material:
Polypropylene
Stainless Steel
Unit Size:
Each
Product Code. 17992032
Brand: Sampling Systems™ 5065B2002
Only null left
Add to basket
Add to basket
These angular beaker attachments allow the cup to be set in specific locations while using the TeleScoop.
  • Telescopic samplers are often used for taking surface samples from rivers, tanks and lakes
  • A wide choice of attachments is available to make the TeleScoop ideally suited for most applications
  • Attachments are interchangeable and simply click into position
Specifications
Sterility Non-sterile
Autoclavable Autoclavable
Capacity (Metric) 2 L
Disposable Reusable
Product Type Swabbing Template
Material Polypropylene