Learn More
Sampling Systems™ Angular Beaker
Product Code. 17982112 Shop All Sampling Systems Products
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1 L
2 L
Material:
Polypropylene
Stainless Steel
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Material unitSize
17982112 1 L Polypropylene Each
17912042 250 mL Polypropylene Each
17902042 500 mL Polypropylene Each
17982032 1 L Stainless Steel Each
17992032 2 L Polypropylene Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17982112 Supplier Sampling Systems™ Supplier No. 5065B1002
Only null left
Add to basket
Add to basket
These angular beaker attachments allow the cup to be set in specific locations while using the TeleScoop.
  • Telescopic samplers are often used for taking surface samples from rivers, tanks and lakes
  • A wide choice of attachments is available to make the TeleScoop ideally suited for most applications
  • Attachments are interchangeable and simply click into position
Specifications
Sterility Non-sterile
Autoclavable Autoclavable
Capacity (Metric) 1 L
Disposable Reusable
Product Type Swabbing Template
Material Polypropylene